Learn More
Creative Biomart Chymopapain
Catalog No. 502084080
Encompass_Preferred
The product was first isolated in 1941 from the crude latex derived from the fruit of Carica papaya by squeezing the green papaya while on the plant prior to harvest. It is an extracellular plant cysteine proteinase similar to papain in specificity.
Catalog No. 50-208-4080 Supplier Creative Biomart Supplier No. THP0029
The product was first isolated in 1941 from the crude latex derived from the fruit of Carica papaya by squeezing the green papaya while on the plant prior to harvest. It is an extracellular plant cysteine proteinase similar to papain in specificity.