Learn More
Cayman Chemical Extracellular Death Factor, Purity: ≥95%, CAS: 960129-66-2, 5 mg
SDP
Catalog No. 502972147
Encompass_Preferred
A linear pentapeptide that communicating cells produce and release, which upon reaching a sufficient concentration activates the cell death pathway in a subset of cells; facilitates mazEF-mediated cell death, significantly reducing population size in E. coli cultures at 2.5 ng/ml
Catalog No. 50-297-2147 Supplier Cayman Chemical Supplier No. 154785 MG
A linear pentapeptide that communicating cells produce and release, which upon reaching a sufficient concentration activates the cell death pathway in a subset of cells; facilitates mazEF-mediated cell death, significantly reducing population size in E. coli cultures at 2.5 ng/ml