Learn More
Sigma Aldrich Fine Chemicals Biosciences Extract-N-AmpTM Blood PCR
Catalog No. 502904152
Encompass
The Extract-N-Amp Blood PCR Kits contain all of the reagents necessary to rapidly extract host genomic DNA from whole blood and amplify targets of interest by PCR (fig. 1). This novel extraction system eliminates the need for any type of purification organic extraction centrifugation heating filtration or alcohol precipitation. The kit also includes a PCR ReadyMix(TM) specially formulated for amplification directly from the extract. This formulation uses an antibody based hot start for specific amplification.
Catalog No. 50-290-4152 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. XNAB21KT
The Extract-N-Amp Blood PCR Kits contain all of the reagents necessary to rapidly extract host genomic DNA from whole blood and amplify targets of interest by PCR (fig. 1). This novel extraction system eliminates the need for any type of purification organic extraction centrifugation heating filtration or alcohol precipitation. The kit also includes a PCR ReadyMix(TM) specially formulated for amplification directly from the extract. This formulation uses an antibody based hot start for specific amplification.