Learn More
Sigma Aldrich Fine Chemicals Biosciences Extract N AmpTM PCR ReadyM
Catalog No. 502914221
Encompass
PCR ReadyMix is intended for use with Sigmas Extract-N-Amp Plant PCR kit and Extract-N-Amp Tissue PCR Kit. All Extract-N-Amp kits include a PCR ReadyMix sufficient for one PCR reaction per extraction. However if additional PCR reactions are required supplemental PCR ReadyMix may be needed.
Catalog No. 50-291-4221 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. E300412ML
PCR ReadyMix is intended for use with Sigmas Extract-N-Amp Plant PCR kit and Extract-N-Amp Tissue PCR Kit. All Extract-N-Amp kits include a PCR ReadyMix sufficient for one PCR reaction per extraction. However if additional PCR reactions are required supplemental PCR ReadyMix may be needed.