Learn More
Sigma Aldrich Fine Chemicals Biosciences Extract N AmpTM Plant PCR
Catalog No. 502905502
Encompass
The Extract-N-Amp Plant PCR Kits contain all the reagents necessary to rapidly extract genomic DNA from plant leaves and amplify targets of interest by PCR. A novel Extraction SOLUTIONon eliminates the need for conventional freezing of plant tissues with liquid nitrogen mechanical disruption organic extraction column purification or precipitation of DNA.
Catalog No. 50-290-5502 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. XNAP21KT
The Extract-N-Amp Plant PCR Kits contain all the reagents necessary to rapidly extract genomic DNA from plant leaves and amplify targets of interest by PCR. A novel Extraction SOLUTIONon eliminates the need for conventional freezing of plant tissues with liquid nitrogen mechanical disruption organic extraction column purification or precipitation of DNA.