Learn More
IW Tremont Co Inc LabExact Extraction thimbles Cellulose Format: 27x80 mm (Inner diameter x Height) pack of 25
SDP
Catalog No. 502237629
Encompass_Preferred
LabExact High quality extraction thimbles available in a variety of dimensions to fit most Soxhlet extraction units. Extraction thimbles are manufactured with high purity cellulose fibers and have a consistent 1.5mm wall thickness. Common applications include air and waste gas analysis, collection of solid particles such as dust, concrete slurry evaluation, analysis of pesticide residues, oil/fat content of solid foods and oil and grease analysis of solid wastes.
This item is not returnable. View return policy
Catalog No. 50-223-7629 Supplier IW Tremont Co Inc Supplier No. LECT5009
LabExact High quality extraction thimbles available in a variety of dimensions to fit most Soxhlet extraction units. Extraction thimbles are manufactured with high purity cellulose fibers and have a consistent 1.5mm wall thickness. Common applications include air and waste gas analysis, collection of solid particles such as dust, concrete slurry evaluation, analysis of pesticide residues, oil/fat content of solid foods and oil and grease analysis of solid wastes.