Learn More
IW Tremont Co Inc LabExact Extraction thimbles Cellulose Format: 33 x 80 mm (Inner diameter x Height) pack of 25
SDP
Catalog No. 502237663
Encompass_Preferred
LabExact High quality extraction thimbles available in a variety of dimensions to fit most Soxhlet extraction units. Extraction thimbles are manufactured with high purity cellulose fibers and have a consistent 1.5mm wall thickness. Common applications include air and waste gas analysis, collection of solid particles such as dust, smoke stack gas monitoring, analysis of pesticide residues, oil/fat content of solid foods, oil and grease analysis of solid wastes and elution for biochemical analysis.
This item is not returnable. View return policy
Catalog No. 50-223-7663 Supplier IW Tremont Co Inc Supplier No. LECT5951
LabExact High quality extraction thimbles available in a variety of dimensions to fit most Soxhlet extraction units. Extraction thimbles are manufactured with high purity cellulose fibers and have a consistent 1.5mm wall thickness. Common applications include air and waste gas analysis, collection of solid particles such as dust, smoke stack gas monitoring, analysis of pesticide residues, oil/fat content of solid foods, oil and grease analysis of solid wastes and elution for biochemical analysis.