Learn More
VWR INTERNATIONAL Extracta DNA prep for PCR, 25 mL
Catalog No. NC1840320
Encompass
Quanta's Extracta sample preparation system is a system that can be scaled to process various amounts of solid tissue in a simple and fast single-tube procedure. DNA may be purified from many tissue types and is suitable for use in sensitive downstream applications such as PCR, qPCR and High Resolution Melt Analysis.
Catalog No. NC1840320 Supplier VWR INTERNATIONAL Supplier No. 97065350
Quanta's Extracta sample preparation system is a system that can be scaled to process various amounts of solid tissue in a simple and fast single-tube procedure. DNA may be purified from many tissue types and is suitable for use in sensitive downstream applications such as PCR, qPCR and High Resolution Melt Analysis.