Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Extractables & Leachables Screening Standard, 18-Component, 1176 Da Mass Range, Dual Ionization

Catalog No. 502038313 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Extractables & Leachables Screening Standard is an 18-component mixture for evaluating and benchmarking high-resolution LC-MS systems. It assesses system preparedness for non-targeted screening and unknown compound analysis. Covering a mass range up to 1176 Da, it supports both positive and negative ionization modes, simplifying screening workflows.

  • Contains 18 common polymer additives
  • Covers a mass range up to 1176 Da
  • Supports positive and negative ionization modes for MS detection
  • Provided as a liquid for direct injection in 2 mL methanol
  • Store at 4°C ± 2°C; shelf life of 12 months
  • Optimized for reversed phase separation with LCMS systems
  • Suitable for quantitative small molecule analysis
  • Concentration tolerance of ± 1%
  • Requires special freight handling for hazardous material

Catalog No. 50-203-8313 Supplier Waters Corp Supplier No. 186008063
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Extractables & Leachables Screening Standard is an 18-component mixture for evaluating and benchmarking high-resolution LC-MS systems. It assesses system preparedness for non-targeted screening and unknown compound analysis. Covering a mass range up to 1176 Da, it supports both positive and negative ionization modes, simplifying screening workflows.

  • Contains 18 common polymer additives
  • Covers a mass range up to 1176 Da
  • Supports positive and negative ionization modes for MS detection
  • Provided as a liquid for direct injection in 2 mL methanol
  • Store at 4°C ± 2°C; shelf life of 12 months
  • Optimized for reversed phase separation with LCMS systems
  • Suitable for quantitative small molecule analysis
  • Concentration tolerance of ± 1%
  • Requires special freight handling for hazardous material

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.