Learn More
Waters Corp WATERS CORPORATION Extraction+ Cartridge Kit, 3 cc, Andrew+/Pipette+ Compatible
Catalog No. 502269312 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Extraction+ 3 cc Cartridge Kit is designed to streamline purchasing and offers compatibility with Andrew+ and Pipette+ systems. It includes essential components for extraction processes.
  • Includes 3 cc Cartridge Adaptor for Extraction+.
  • Comes with μ12mm Vial Collection Rack.
  • Contains a pack of 3 cc Sealing Caps (x24).
  • Features a Collection Rack Domino.
  • Compatible with OneLab software.
  • Functions as a Connected Device.
This item is not returnable. View return policy
Catalog No. 50-226-9312 Supplier Waters Corp Supplier No. 176005204
The Extraction+ 3 cc Cartridge Kit is designed to streamline purchasing and offers compatibility with Andrew+ and Pipette+ systems. It includes essential components for extraction processes.
  • Includes 3 cc Cartridge Adaptor for Extraction+.
  • Comes with μ12mm Vial Collection Rack.
  • Contains a pack of 3 cc Sealing Caps (x24).
  • Features a Collection Rack Domino.
  • Compatible with OneLab software.
  • Functions as a Connected Device.