Learn More
Waters Corp Waters Cartridge Kit, 3 cc, for Extraction+
Catalog No. 502269313 Shop All Waters Corporation Products
Encompass_Preferred
The Waters Extraction+ 3 cc Cartridge Kit provides essential components for laboratory extraction processes. It is designed for use with Andrew+ and Pipette+ systems and is compatible with OneLab software.
  • Includes a 3 cc cartridge adaptor for Extraction+ systems.
  • Features a 12 mm vial collection rack.
  • Comes with a pack of 24, 3 cc sealing caps.
  • Contains a collection rack domino.
  • Compatible with Andrew+ and Pipette+ systems.
  • Integrates with OneLab software for seamless operation.
  • Provided as a single package.
This item is not returnable. View return policy
Catalog No. 50-226-9313 Supplier Waters Corp Supplier No. 176005205
The Waters Extraction+ 3 cc Cartridge Kit provides essential components for laboratory extraction processes. It is designed for use with Andrew+ and Pipette+ systems and is compatible with OneLab software.
  • Includes a 3 cc cartridge adaptor for Extraction+ systems.
  • Features a 12 mm vial collection rack.
  • Comes with a pack of 24, 3 cc sealing caps.
  • Contains a collection rack domino.
  • Compatible with Andrew+ and Pipette+ systems.
  • Integrates with OneLab software for seamless operation.
  • Provided as a single package.