Learn More
Chemglass Life Sciences Micro Size SOXHLET Extractor, Complete
SDP
Catalog No. CG137401
Encompass_Preferred
Micro Size SOXHLET Extractor, Complete. Micro size Soxhlet extractor for use with 10 x 50mm paper or glass thimbles. Extractor body has a 19/22 top inner joint and a 14/20 lower inner joint. Complete apparatus consists of the extractor body, condenser, and 25mL round bottom flask. Glass thimbles must be ordered separately.
Catalog No. CG137401 Supplier Chemglass Life Sciences Supplier No. CG137401
Micro Size SOXHLET Extractor, Complete. Micro size Soxhlet extractor for use with 10 x 50mm paper or glass thimbles. Extractor body has a 19/22 top inner joint and a 14/20 lower inner joint. Complete apparatus consists of the extractor body, condenser, and 25mL round bottom flask. Glass thimbles must be ordered separately.