Learn More
Chemglass Life Sciences Small Extraction Apparatus Complete, Soxhlet, 34/45, 24/40
SDP
Catalog No. CG136801
Encompass_Preferred
Extraction Apparatus, Soxhlet, Small, 34/45, 24/40. Soxhlet type extraction apparatus used for continuous extraction of solids with a suitable solvent. Extractor uses paper or CG-1372 glass thimbles. Condenser has an enlarged uptake tube to improve condensing capacity. Complete apparatus consists of an Allihn condenser, extractor and flat bottom flask.
Catalog No. CG136801 Supplier Chemglass Life Sciences Supplier No. CG136801
Extraction Apparatus, Soxhlet, Small, 34/45, 24/40. Soxhlet type extraction apparatus used for continuous extraction of solids with a suitable solvent. Extractor uses paper or CG-1372 glass thimbles. Condenser has an enlarged uptake tube to improve condensing capacity. Complete apparatus consists of an Allihn condenser, extractor and flat bottom flask.