Learn More
Waters Corp Waters Amino Acid Analysis Kit, C18, 2.5 um, for UHPLC, HPLC
Supplier:  Waters Corp 176005152
Encompass_Preferred
This kit provides all the necessary materials to run AccQ·Tag Ultra chemistries on UHPLC and HPLC systems, offering a complete solution for amino acid analysis.
  • Provides materials for 250 analyses.
  • Includes C18 2.5 μm 4.6 × 150 mm column.
  • Contains Eluent A concentrate 1 L and Eluent B 1 L.
  • Supplied with amino acid standard, hydrolysate 10 × 1 mL.
  • Includes total recovery vials with caps 3 × 100/pk.
  • Supports UPLC/UHPLC/HPLC separation modes.
  • Utilizes UV detection method.
  • Suitable for UHPLC and HPLC instruments.
  • Shipped at room temperature.
  • Designed for amino acid applications.
Catalog No. 50-226-9308