Learn More
Abcam E x traction Buffer 5 x PTR, 50ML
Catalog No. NC1558185
Encompass_Preferred
Extraction buffer 5X PTR is formulated for preparing cell or tissue extracts for use in immunoassays, with added phosphatase inhibitors and protease inhibitor aprotinin. - Cited in over 20 publications - Phosphatase and protease inhibitors – contains inhibitors to preserve sample integrity - Easy to use – dilute to 1X with distilled water before use, or use with extraction enhancer buffer 50X for enhanced extraction - Compatible with a variety of sample types
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1558185 Supplier Abcam Supplier No. AB19397050ML
Add to Cart
Add to Cart
Extraction buffer 5X PTR is formulated for preparing cell or tissue extracts for use in immunoassays, with added phosphatase inhibitors and protease inhibitor aprotinin. - Cited in over 20 publications - Phosphatase and protease inhibitors – contains inhibitors to preserve sample integrity - Easy to use – dilute to 1X with distilled water before use, or use with extraction enhancer buffer 50X for enhanced extraction - Compatible with a variety of sample types
The product is subject to the following: Abcam Restricted Use Statement