Learn More
Waters Corp Waters Extraction Cone, 1.17 mm Aperture
Catalog No. 50466685 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Extraction Cone is designed with a 1.17 mm aperture. This spare part is essential for specific extraction processes in laboratory settings.
  • Features a 1.17 mm aperture
  • Comes in a single pack
  • Can be replaced as required
Catalog No. 50-466-685 Supplier Waters Corp Supplier No. 700005998
The Extraction Cone is designed with a 1.17 mm aperture. This spare part is essential for specific extraction processes in laboratory settings.
  • Features a 1.17 mm aperture
  • Comes in a single pack
  • Can be replaced as required