Learn More
Velp™ Extraction Cup
SDP
Catalog No. 11392031
Change view
Click to view available options
Product Type:
Extraction Cup
Catalog No. Product Type
11-392-031 Extraction Cup
1 options
This item is not returnable. View return policy
Catalog No. 11-392-031 Supplier Velp™ Supplier No. A00000142
Add to Cart
Add to Cart
This item is not returnable. View return policy
Velp™ Extraction Cup for use with SER 148 Series Solvent Extractors.
  • For use with SER 148 Series Solvent Extractor
Specifications
Quantity 6 Pk.
Material Glass
For Use With (Equipment) SER 148 Series Solvent Extractors
Product Type Extraction Cup
Sterility Sterile