Learn More
Velp™ Scientific Extraction Cups for Solvent Extractor
SDP
Catalog No. 11392090
Change view
Click to view available options
Diameter (Metric):
48 mm
65 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
11-392-090 48 mm
11-392-089 65 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 11-392-090 Supplier Velp™ Scientific Supplier No. A00000303
Add to Cart
Add to Cart
This item is not returnable. View return policy
Extraction cup for use with the SER158/3 Solvent Extractor.
  • Various sizes of cups for solvent extraction
Specifications
Product Type Extraction Cup
For Use With (Equipment) SER158/3 Solvent Extractor
Quantity 3 Cups
Diameter (Metric) 48 mm
Height (Metric) 120 mm