Learn More
GENEPATH DIAGNOSTICS INC Extraction-free viral medium (VCM) enables direct-to-PCR workflows for Influenza A, Influenza B and SARS-CoV2 assays without the need for RNA extraction (RUO)
Catalog No. NC3949361
Encompass_Preferred
Supports preservation and stabilization of viral nucleic acids along with direct addition into compatible PCR mixes (without the need for additional sample preparation or nucleic acid extraction). Suitable for molecular detection of SARS-CoV-2, Influenza A/B, and others. Sample Type: NP/OP swabs, saliva swabs. (RUO)
Catalog No. NC3949361 Supplier GENEPATH DIAGNOSTICS INC Supplier No. 10510200100
Supports preservation and stabilization of viral nucleic acids along with direct addition into compatible PCR mixes (without the need for additional sample preparation or nucleic acid extraction). Suitable for molecular detection of SARS-CoV-2, Influenza A/B, and others. Sample Type: NP/OP swabs, saliva swabs. (RUO)