Learn More
Waters Corp Waters Extraction+ Pump and Manifold, Fully Automated, Andrew+ and Pipette+ Compatible
Catalog No. 502269411 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Extraction+ Pump and Manifold is a connected device designed for new or existing Andrew+ customers. It offers a plug-and-play solution for extraction methods that can be fully automated with the Andrew+ robot. This device is compatible with μElution, macro elution plates, and 1 cc, 3 cc, and 6 cc cartridges.
  • Software-controlled pump enables automated pressure profiling control.
  • Machine-readable vacuum manifold base allows for full automation of extraction methods with the Andrew+ robot.
  • Ensures continuous waste flow-through.
  • Compatible with Andrew+ and Pipette+ systems.
  • Works seamlessly with OneLab software.
This item is not returnable. View return policy
Catalog No. 50-226-9411 Supplier Waters Corp Supplier No. 186010524
Add to Cart
Add to Cart
Request a Quote
The Extraction+ Pump and Manifold is a connected device designed for new or existing Andrew+ customers. It offers a plug-and-play solution for extraction methods that can be fully automated with the Andrew+ robot. This device is compatible with μElution, macro elution plates, and 1 cc, 3 cc, and 6 cc cartridges.
  • Software-controlled pump enables automated pressure profiling control.
  • Machine-readable vacuum manifold base allows for full automation of extraction methods with the Andrew+ robot.
  • Ensures continuous waste flow-through.
  • Compatible with Andrew+ and Pipette+ systems.
  • Works seamlessly with OneLab software.