Learn More
Waters Corp Waters Extraction Plate Manifold Kit, Vacuum, 96-well Plate, μElution Spacer
Catalog No. 50786012 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This vacuum manifold is designed for vacuum extraction with 96-well plate products. The kit includes a spacer specifically for use with μElution plates.
  • Designed for vacuum extraction with 96-well plate products.
  • Includes a spacer for use with μElution plates.
  • Contains reservoir trays.
  • Contains sealing caps.
  • Contains 2 mL sample collection plates.
Catalog No. 50-786-012 Supplier Waters Corp Supplier No. WAT097946
This vacuum manifold is designed for vacuum extraction with 96-well plate products. The kit includes a spacer specifically for use with μElution plates.
  • Designed for vacuum extraction with 96-well plate products.
  • Includes a spacer for use with μElution plates.
  • Contains reservoir trays.
  • Contains sealing caps.
  • Contains 2 mL sample collection plates.