Learn More
Waters Corp Waters Extraction Plate Manifold Kit, for 96-Well Plates, Vacuum, with μElution Spacer
Catalog No. 50786011 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The vacuum manifold for 96-well plates is designed for the vacuum extraction of all 96-well plate products, including Oasis, Ostro, and Sep-Pak. This kit also contains a spacer necessary for use with μElution plates.
  • Ideally suited for vacuum extraction with 96-well plate products.
  • Works with all 96-well plate products (Oasis, Ostro, Sep-Pak).
  • Includes a spacer for μElution plates.
  • Extraction plate manifold.
  • Reservoir trays.
  • Sealing caps.
  • 1 mL sample collection plates.
Catalog No. 50-786-011 Supplier Waters Corp Supplier No. WAT097945
The vacuum manifold for 96-well plates is designed for the vacuum extraction of all 96-well plate products, including Oasis, Ostro, and Sep-Pak. This kit also contains a spacer necessary for use with μElution plates.
  • Ideally suited for vacuum extraction with 96-well plate products.
  • Works with all 96-well plate products (Oasis, Ostro, Sep-Pak).
  • Includes a spacer for μElution plates.
  • Extraction plate manifold.
  • Reservoir trays.
  • Sealing caps.
  • 1 mL sample collection plates.