Learn More
Waters Corp Waters Corporation Extraction Plate Manifold Kit, for 96-Well Plates, Vacuum Extraction
Catalog No. 50818755 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Vacuum Manifold for 96-well plates is designed for vacuum extraction with all 96-well plate products, including Oasis, Ostro, and Sep-Pak. The kit contains an extraction plate manifold, reservoir trays, sealing caps, and 350 μL sample collection plates. It also includes a spacer necessary for use with μElution plates.
  • Ideally suited for vacuum extraction of all 96-well plate products including Oasis, Ostro, and Sep-Pak.
  • Includes a spacer required for use with μElution plates.
  • Kit includes an extraction plate manifold, reservoir trays, sealing caps, and 350 μL sample collection plates.
Catalog No. 50-818-755 Supplier Waters Corp Supplier No. WAT097944
The Vacuum Manifold for 96-well plates is designed for vacuum extraction with all 96-well plate products, including Oasis, Ostro, and Sep-Pak. The kit contains an extraction plate manifold, reservoir trays, sealing caps, and 350 μL sample collection plates. It also includes a spacer necessary for use with μElution plates.
  • Ideally suited for vacuum extraction of all 96-well plate products including Oasis, Ostro, and Sep-Pak.
  • Includes a spacer required for use with μElution plates.
  • Kit includes an extraction plate manifold, reservoir trays, sealing caps, and 350 μL sample collection plates.