Learn More
Waters Corp Waters Collection Rack Domino, Automated SPE, Andrew+ Robot Compatible
Catalog No. 502269410 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Collection Rack Domino is designed for fully automated Solid Phase Extraction (SPE) workflows. It facilitates automated movements of consumables to and from the Extraction+ manifold base using an Andrew+ robot equipped with a Microplate Gripper Tool, and is compatible with an expanding range of Collection Racks.
  • Enables fully automated SPE workflows.
  • Allows automated consumable movements.
  • Compatible with an expanding range of collection racks.
  • Designed for use with the Andrew+ robot and Microplate Gripper Tool.
This item is not returnable. View return policy
Catalog No. 50-226-9410 Supplier Waters Corp Supplier No. 186010523
The Collection Rack Domino is designed for fully automated Solid Phase Extraction (SPE) workflows. It facilitates automated movements of consumables to and from the Extraction+ manifold base using an Andrew+ robot equipped with a Microplate Gripper Tool, and is compatible with an expanding range of Collection Racks.
  • Enables fully automated SPE workflows.
  • Allows automated consumable movements.
  • Compatible with an expanding range of collection racks.
  • Designed for use with the Andrew+ robot and Microplate Gripper Tool.