Learn More
Enzo Life Sciences Extraction reagent 1 (5X), (10ml)
SDP
Catalog No. 502006821
Encompass_Preferred
Concentrated lysis buffer for preparation of cell and tissue extracts. Formulation: Liquid. TRIS buffer containing detergents. Long Term Storage: +4°C.
Catalog No. 50-200-6821 Supplier Enzo Life Sciences Supplier No. ADI801526
Concentrated lysis buffer for preparation of cell and tissue extracts. Formulation: Liquid. TRIS buffer containing detergents. Long Term Storage: +4°C.