Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Extraction Solvent, 4mL/L Hydrochloric Acid in Reagent Alcohol, LabChem™

Catalog No. LC141804 Shop All LabChem Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC141804 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC141804 Supplier LabChem, Inc. Supplier No. LC141804
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Extraction Solvent
Color Colorless
Name Note 4mL/L Hydrochloric Acid in Reagent Alcohol
CAS Max % 89.6, 5, 5, 0.4
CAS 67-63-0, 67-56-1, 7647-01-0
Linear Formula HCl
UN Number UN1993
Solubility Information Soluble in water
Formula Weight 36.46
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide; May release flammable gases
Quantity 4 L
Physical Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.