Learn More
Ace Glass, Inc. Soxhlet extraction system, 125mL, consists of condenser, extractor and flask, use 27mm ID thimbles (6811-08, -14)
SDP
Catalog No. 50973396
Encompass_Preferred
125mL Soxhlet extraction system featuring condenser, extractor, and flask. The ACE design permits greater condensing capacity as cycling rates may be doubled over conventional style extractors. Use with ACE 6811 27mm I.D. thimbles.
Catalog No. 50-973-396 Supplier Ace Glass, Inc. Supplier No. 671612
125mL Soxhlet extraction system featuring condenser, extractor, and flask. The ACE design permits greater condensing capacity as cycling rates may be doubled over conventional style extractors. Use with ACE 6811 27mm I.D. thimbles.