Learn More
Ace Glass, Inc. Soxhlet extraction system, 300mL, consists of condenser, extractor and flask, use 33mm ID thimble (6811-22)
SDP
Catalog No. 50978872
Encompass_Preferred
300mL Soxhlet extraction system featuring condenser, extractor, and flask. The ACE design permits greater condensing capacity as cycling rates may be doubled over conventional style extractors. Use with ACE 6811 33mm I.D. thimble.
Catalog No. 50-978-872 Supplier Ace Glass, Inc. Supplier No. 671618
300mL Soxhlet extraction system featuring condenser, extractor, and flask. The ACE design permits greater condensing capacity as cycling rates may be doubled over conventional style extractors. Use with ACE 6811 33mm I.D. thimble.