Learn More
Ace Glass, Inc. Soxhlet extraction system, 300mL, consists of condenser, extractor and flask, use 40mm ID thimble (6811-24)
SDP
Catalog No. 50973398
Encompass_Preferred
300mL Soxhlet extraction system featuring condenser, extractor, and flask. The ACE design permits greater condensing capacity as cycling rates may be doubled over conventional style extractors. Use with ACE 6811 40mm I.D. thimble.
Catalog No. 50-973-398 Supplier Ace Glass, Inc. Supplier No. 671620
300mL Soxhlet extraction system featuring condenser, extractor, and flask. The ACE design permits greater condensing capacity as cycling rates may be doubled over conventional style extractors. Use with ACE 6811 40mm I.D. thimble.