Learn More
Velp™ Scientific Extraction Thimble Holder
SDP
Catalog No. 11392079
Change view
Click to view available options
For Use With (Equipment):
SER158/3 Solvent Extractor
Catalog No. For Use With (Equipment)
11-392-079 SER158/3 Solvent Extractor
1 options
This item is not returnable. View return policy
Catalog No. 11-392-079 Supplier Velp™ Scientific Supplier No. A00000292
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thimble holder for extractions.
  • Holds thimbles that are 40 mm in diameter
Specifications
Product Type Thimble Holder
For Use With (Equipment) SER158/3 Solvent Extractor
Diameter (Metric) 40 mm