Learn More
Ace Glass, Inc. EXTRACTOR BODY MICRO SOXHLET EXTRACTOR BODY MICRO SOXHLET
SDP
Catalog No. 50973418
Encompass_Preferred
Extractor body for ACE micro Soxhlet extractor (50973419). Soxhlet extractor is designed for small chemical applications. Improved design permits greater condensing capacity.
Catalog No. 50-973-418 Supplier Ace Glass, Inc. Supplier No. 677604
Extractor body for ACE micro Soxhlet extractor (50973419). Soxhlet extractor is designed for small chemical applications. Improved design permits greater condensing capacity.