Learn More
Chemglass Life Sciences Extractor, Liquid-Liquid, Continous, 45/50, 24/40
SDP
Catalog No. 50873060
Encompass_Preferred
Extractor, Liquid-Liquid, Continuous, 45/50, 24/40. Round bottom design has the return tube sealed into the bottom of the extractor for more efficient extractions. For samples of 1 liter plus approximately 200mL of extracting solvent. Extractor has a 45/50 top outer joint and a 24/40 inner joint on the uptake tube. Flask and condenser must be ordered separately.
Catalog No. 50-873-060 Supplier Chemglass Life Sciences Supplier No. CG136601
Extractor, Liquid-Liquid, Continuous, 45/50, 24/40. Round bottom design has the return tube sealed into the bottom of the extractor for more efficient extractions. For samples of 1 liter plus approximately 200mL of extracting solvent. Extractor has a 45/50 top outer joint and a 24/40 inner joint on the uptake tube. Flask and condenser must be ordered separately.