Learn More
Med Vet International Miltex Saafield Comedone Extractor, 5.75in., Double-Ended, 5.1mm Spoon End, 2.8mm Acne Lancet End
Catalog No. 502829679
Encompass_Preferred
The Miltex 33-204 Saafield comedone extractor is made of OR grade German stainless steel and measures 5.75" in length. It is double-ended with a 5.1mm spoon end and a 2.8mm acne lancet end.
Catalog No. 50-282-9679 Supplier Med Vet International Supplier No. MIL33204
The Miltex 33-204 Saafield comedone extractor is made of OR grade German stainless steel and measures 5.75" in length. It is double-ended with a 5.1mm spoon end and a 2.8mm acne lancet end.