Learn More
Wilmad Labglass EXTRACTOR SXHLT 40MM ID
Catalog No. 50804454
Encompass_Preferred
Extractor, Sohxlet. Extractor Capacity: 85mL; Upper Joint Size: [ST] 45/50; Lower Joint Size: 24/40. Replacement component For LG-6900-104 Soxhlet Extraction Apparatus.
Catalog No. 50-804-454 Supplier Wilmad Labglass Supplier No. LG6900124
Extractor, Sohxlet. Extractor Capacity: 85mL; Upper Joint Size: [ST] 45/50; Lower Joint Size: 24/40. Replacement component For LG-6900-104 Soxhlet Extraction Apparatus.