Learn More
Wilmad Labglass EXTRACTOR SXHLT 50MM ID
Catalog No. 50803773
Encompass_Preferred
Extractor, Sohxlet. Extractor Capacity: 200mL; Upper Joint Size: [ST] 55/50; Lower Joint Size: 24/40. Replacement component For LG-6900-108 Soxhlet Extraction Apparatus.
Catalog No. 50-803-773 Supplier Wilmad Labglass Supplier No. LG6900128
Extractor, Sohxlet. Extractor Capacity: 200mL; Upper Joint Size: [ST] 55/50; Lower Joint Size: 24/40. Replacement component For LG-6900-108 Soxhlet Extraction Apparatus.