Learn More
MSC Armor Guys Kyorene Pro Extraflex Cut Resistant Glove, ANSI A4, Graphene Liner, Size S, 12/Pk.
Catalog No. 5000688394
Encompass
These cut-resistant gloves provide advanced protection and dexterity for high-risk environments. Constructed with an 18g graphene liner, they deliver high-performance cut resistance while maintaining comfort and tactile sensitivity. The gloves feature a microfoam nitrile coating that enhances grip in various conditions.
  • ANSI A4 cut resistance level for superior protection
  • Lightweight 18g graphene liner design
  • Microfoam nitrile coating for enhanced grip
  • Size S
  • Pack of 12 pairs
Catalog No. 50-006-88394 Supplier MSC Supplier No. 96751532
Add to Cart
Add to Cart
These cut-resistant gloves provide advanced protection and dexterity for high-risk environments. Constructed with an 18g graphene liner, they deliver high-performance cut resistance while maintaining comfort and tactile sensitivity. The gloves feature a microfoam nitrile coating that enhances grip in various conditions.
  • ANSI A4 cut resistance level for superior protection
  • Lightweight 18g graphene liner design
  • Microfoam nitrile coating for enhanced grip
  • Size S
  • Pack of 12 pairs