Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Extran™ AP 41

Catalog No. M1075709025 Shop All MilliporeSigma Products
Change view
Click to view available options
Volume (Metric):
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
M1075709025 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1075709025 Supplier MilliporeSigma™ Supplier No. 1075709025

missing translation for 'validMainframeMsg'

Recommended for use on glass, stainless steel, porcelain, plastic, rubber, and fiberglass. MilliporeSigma™ Extran™ AP 41 is an enzymatic powdered cleaner designed to remove mucus, saliva, blood, and protein residues. Dissolve 3g/L and use at 60°C

The magnesium concentration is measured semiquantitatively by visual comparison of the colour of the measurement solution with the colour fields of a colour card.

TRUSTED_SUSTAINABILITY

Specifications

Form Powdered Solid
pH 11.4 (3 g/L aq. soln., 20°C)
Appearance White
Composition Enzymes, phosphates, and alkali salts
Container Type Plastic drum
Density 750 kg/m3
For Use With (Application) Remove mucus, saliva, blood, and protein residues
Volume (Metric) 25 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.