Learn More
MilliporeSigma™ Extran™ AP 41
Recommended for use on glass, stainless steel, porcelain, plastic, rubber, and fiberglass. MilliporeSigma™ Extran™ AP 41 is an enzymatic powdered cleaner designed to remove mucus, saliva, blood, and protein residues. Dissolve 3g/L and use at 60°C
Supplier: MilliporeSigma™ 1075702000
Description
The Carbonate Hardness Test determines carbonate hardness as a measure of the buffering capacity of water which can also be expressed as acid-binding capacity or total alkalinity.