Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Extran™ MN 01

Catalog No. MEX09955 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MEX09955 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MEX09955 Supplier MilliporeSigma™ Supplier No. EX09955

Dissolves easily, even in hard water. MilliporeSigma™ Extran™ MN 01 is a phosphate-free powdered cleaner formulated with anionic surfactants. This concentrated universal cleaner can be used for heavy-duty manual cleaning of glass, quartz, metal, plast

TRUSTED_SUSTAINABILITY

Specifications

Form Powdered Solid
pH 7 to 9 (1%, aq. soln.)
Appearance Off-White
Composition Anionic surfactants and auxiliary agents
Container Type Plastic drum
For Use With (Application) Heavy-duty manual washing
Quantity 10 kg
Volume (Metric) 10 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.