Learn More
Cole-Parmer EH-200-2-L 2-Well Macro-Kjeldahl Extraction Heater, 800 mL, UK; 230 VAC
Catalog No. 501960379 Shop All Cole-Parmer Products
Encompass_Preferred
These multiposition extraction heaters let you run several projects simultaneously by using either two or six individually controlled heating mantles. Each mantle has an independent heater with control dial, on/off switch, and indicator light, and operates to a maximum element temperature of 800C. A quartz heating element is housed in a thermally insulated cartridge for easy replacement, extending the life of the heater. Heaters are supplied with back-mounted brackets to hold support rods up to 1/2" (12.7 mm) in diameter. The lower part of the unit containing the control equipment is safely separated from the upper heating elelments by a well-ventilated air space. Units are double fused on the supply line and neutral for extra safety. Power cord is NOT included. Unit must be hard-wired.
Catalog No. 50-196-0379 Supplier Cole-Parmer Supplier No. 0307508
These multiposition extraction heaters let you run several projects simultaneously by using either two or six individually controlled heating mantles. Each mantle has an independent heater with control dial, on/off switch, and indicator light, and operates to a maximum element temperature of 800C. A quartz heating element is housed in a thermally insulated cartridge for easy replacement, extending the life of the heater. Heaters are supplied with back-mounted brackets to hold support rods up to 1/2" (12.7 mm) in diameter. The lower part of the unit containing the control equipment is safely separated from the upper heating elelments by a well-ventilated air space. Units are double fused on the supply line and neutral for extra safety. Power cord is NOT included. Unit must be hard-wired.