Learn More
MilliporeSigma™ Extrelut™ NT Packing
Catalog No. M1150930001 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Bulk Material
Refill Pack
For Use With (Equipment):
EXtrelut NT1, NT3 and NT20
EXtrelut NT20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
M1150930001 Refill Pack EXtrelut NT20
M1150921000 Bulk Material EXtrelut NT1, NT3 and NT20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1150930001 Supplier MilliporeSigma™ Supplier No. 1150930001
Add to Cart
Add to Cart
Use in the production of large extraction columns. MilliporeSigma EXtrelut™ NT Packing is made of pure Kieselguhr (CAS 68855-54-9, pH 10) and is available in a bulk pack for use with all Extrelut™ NT columns or pre-portioned refill packs specif
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) EXtrelut NT20
Type Refill Pack