Learn More
Thomas Scientific eXTReme FoilSeal Sealing Foils, Non-Sterile, 100/pk
SDP
Catalog No. NC1743117
Encompass
eXTReme FoilSeal is an aluminum foil seal that is excellent for chemical resistance and superior performance in ultra-low temperatures.
Catalog No. NC1743117 Supplier Thomas Scientific Supplier No. 1180W00
eXTReme FoilSeal is an aluminum foil seal that is excellent for chemical resistance and superior performance in ultra-low temperatures.