Learn More
Refrigiwear Inc. EXTREME FREEZER GLOVE
SDP
Catalog No. NC2337320
Encompass_Preferred
Built for hard work in the coldest temperatures, you can put the Extreme Freezer Gloves through the ringer time and time again. With impact protection and just about the most durable construction you'll find, these insulated work gloves protect your hands from hazards on the job as well as harsh -30°F conditions. The knitted fabric back with a synthetic leather palm and silicone grip pattern on the palm keeps your hands warm and your grip secure. Even the strongest winds won't cut through the 200g insulation in these tough impact protection gloves. The entire Extreme Collection, was designed for comfort, style, and the ultimate protection. Comfort Rating: -30°F 200g AirBlaze® insulation laminated to a tricot lining Knitted fabric back with synthetic leather palm. Stretch neoprene cuff Pull tab with rubber ID tag on wrist Meets EN 511:2006 Protective Gloves Against Cold: 2 | 4 | 0 Meets EN 388:2016 Protective Gloves Against Mechanical Risks: 3 | 3 | 3 | 2 | X    
Catalog No. NC2337320 Supplier Refrigiwear Inc. Supplier No. 0679
Built for hard work in the coldest temperatures, you can put the Extreme Freezer Gloves through the ringer time and time again. With impact protection and just about the most durable construction you'll find, these insulated work gloves protect your hands from hazards on the job as well as harsh -30°F conditions. The knitted fabric back with a synthetic leather palm and silicone grip pattern on the palm keeps your hands warm and your grip secure. Even the strongest winds won't cut through the 200g insulation in these tough impact protection gloves. The entire Extreme Collection, was designed for comfort, style, and the ultimate protection. Comfort Rating: -30°F 200g AirBlaze® insulation laminated to a tricot lining Knitted fabric back with synthetic leather palm. Stretch neoprene cuff Pull tab with rubber ID tag on wrist Meets EN 511:2006 Protective Gloves Against Cold: 2 | 4 | 0 Meets EN 388:2016 Protective Gloves Against Mechanical Risks: 3 | 3 | 3 | 2 | X    