Learn More
Refrigiwear Inc. Extreme Freezer Glove, Black, Medium
SDP
Catalog No. NC1083860
Encompass_Preferred
The Extreme Freezer Glove features impact protection, a reinforced thumb crotch, Ergo Fit increased durability with ergonomic design to reduce hand fatigue, wrap around padding on fingers, and shock absorbing pads on the palms and fingers to protect hands from bangs and bruises. Strategically placed pads at contact points keep hands warm when holding cold items. They also feature 100g Thinsulate plus foam insulation with tricot lining, a knitted fabric back with synthetic leather palm, pre-curved ergonomic fit, silicone grip pattern on palm, EVA palm pads, rolled fingertips, silver reflective strip on knuckles, a stretch neoprene cuff, and a pull tab with rubber ID tag on wrist. Available in black, sizes M - 2XL.
This item is not returnable. View return policy
Catalog No. NC1083860 Supplier Refrigiwear Inc. Supplier No. 0679RBLKMED
The Extreme Freezer Glove features impact protection, a reinforced thumb crotch, Ergo Fit increased durability with ergonomic design to reduce hand fatigue, wrap around padding on fingers, and shock absorbing pads on the palms and fingers to protect hands from bangs and bruises. Strategically placed pads at contact points keep hands warm when holding cold items. They also feature 100g Thinsulate plus foam insulation with tricot lining, a knitted fabric back with synthetic leather palm, pre-curved ergonomic fit, silicone grip pattern on palm, EVA palm pads, rolled fingertips, silver reflective strip on knuckles, a stretch neoprene cuff, and a pull tab with rubber ID tag on wrist. Available in black, sizes M - 2XL.