Learn More
Fisherbrand™ Extreme Precision Fine Knife Tip Probe
Catalog No. 12000154 Shop All Fisherbrand Products
Change view
Click to view available options
Tip Style:
Fine knife tip
Catalog No. Tip Style
12-000-154 Fine knife tip
1 options
Not for internal use
Catalog No. 12-000-154
Add to Cart
Add to Cart
Not for internal use
With standard handle.
  • Stainless-steel
  • 10 mil tip and 3 in. L
Note: Minimum order quantity is 6.
Specifications
Description Extreme precision probe
Material Stainless Steel
Tip Style Fine knife tip