Learn More
Research Products International Corp eXTReme Seal, Clear, Non-sterile, 100/pk
SDP
Catalog No. 501978146
Encompass
eXTReme is ideal for PCR, light sensitive samples, robotics, coldroom and longterm storage. Offers universal fit for use with all plate types, minimizes sample evaporation and reduces well-to-well cross contamination. Specially formulated adhesive provides excellent seal for all plates. Removes cleanly from plate with no adhesive residue and does not tear upon removal. Smaller dimensions and chamfered corners result in no plate overhang for robotics. eXTReme is a Trademark of Excel Scientific, Inc.
PCR and longterm storage
RNase, DNase free
Withstands -20°C to +120°C
Pierceable, autoclavable
Catalog No. 50-197-8146 Supplier Research Products International Corp Supplier No. 202560
eXTReme is ideal for PCR, light sensitive samples, robotics, coldroom and longterm storage. Offers universal fit for use with all plate types, minimizes sample evaporation and reduces well-to-well cross contamination. Specially formulated adhesive provides excellent seal for all plates. Removes cleanly from plate with no adhesive residue and does not tear upon removal. Smaller dimensions and chamfered corners result in no plate overhang for robotics. eXTReme is a Trademark of Excel Scientific, Inc.
PCR and longterm storage
RNase, DNase free
Withstands -20°C to +120°C
Pierceable, autoclavable