Learn More
MOLECULAR CLONING LABORATORIES LLC Extreme Thermostable SSB (ET SSB), 125ug, Concentration 500 ug/ml
Catalog No. NC2611642
Encompass
Extreme Thermostable SSB is a single-stranded DNA binding protein isolated from a hyperthermophilic microorganism, which remains fully active after incubation at 95°C for 60 minutes.
Catalog No. NC2611642 Supplier MOLECULAR CLONING LABORATORIES LLC Supplier No. ETSSB100
Extreme Thermostable SSB is a single-stranded DNA binding protein isolated from a hyperthermophilic microorganism, which remains fully active after incubation at 95°C for 60 minutes.