Learn More
MSC Honeywell Safety TurboLite™ EXTREME Self-Retracting Lifeline, Sharp Edge Twin Leg, 9 ft, 420 lb Capacity, 1 ea
Catalog No. 5000509211
Encompass
The TurboLite™ EXTREME sharp edge twin leg personal fall limiter is designed for fall protection in short range applications. This self-retracting lifeline features a durable construction and is engineered for use in environments where sharp edges may be present, providing a reliable fall arrest system for users.
  • Features a 420 lb load capacity
  • Twin leg design for continuous fall protection
  • Equipped with a galvanized steel line
  • Constructed with an aluminum hub, anodized aluminum connector, ballistic nylon cover, high impact nylon housing, stainless steel mechanism, and steel hook hardware
  • Offers 3,000 lb tensile strength
  • Meets ANSI A10.32 and ANSI Z359.14 Class B specifications
  • Suitable for a wide range of short range applications
Catalog No. 50-005-09211 Supplier MSC Supplier No. 41773581
The TurboLite™ EXTREME sharp edge twin leg personal fall limiter is designed for fall protection in short range applications. This self-retracting lifeline features a durable construction and is engineered for use in environments where sharp edges may be present, providing a reliable fall arrest system for users.
  • Features a 420 lb load capacity
  • Twin leg design for continuous fall protection
  • Equipped with a galvanized steel line
  • Constructed with an aluminum hub, anodized aluminum connector, ballistic nylon cover, high impact nylon housing, stainless steel mechanism, and steel hook hardware
  • Offers 3,000 lb tensile strength
  • Meets ANSI A10.32 and ANSI Z359.14 Class B specifications
  • Suitable for a wide range of short range applications