Learn More
Sigma Aldrich Fine Chemicals Biosciences eXTRemeTM Universal fit PC
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z742420100EA
Encompass
XTR films offer high performance under extreme conditions. A 2 mil polyester film with a 1.65 mil layer of strong acrylic adhesive. eXTReme Seal films remove cleanly from plate surfaces without adhesive stringing residue or contamination. Each sealing films measures 78.3 x 140.5 mm and offers sufficient sealing area for all plate types (e.g. flat top and raised-rim). Chamfered corners result in no plate overhang for robotics applications. features two end-tabs with perforations. Length between perforations with end tabs removed is 119 mm.
Catalog No. 50-290-0559