Learn More
Grainger Parker Flare Connector, Brass, 1/2 in. to 5/8 in., 10/Pk.
Catalog No. 502784613
Encompass_Preferred
This brass extruded reducer coupling features a 45° flare design, providing a reliable metal-to-metal seal for high-pressure fluid and gas handling applications. Engineered for safety and dependability, this connector is designed for use with copper, aluminum, brass, and welded steel hydraulic tubing that can be flared. It offers an efficient alternative to soldered joints in environments where open flames are impractical.
  • Constructed from durable brass
  • Designed to meet SAE J512 and SAE J513 standards
  • Suitable for use with water, air, inert and non-combustible gases, flammable liquids, and Freon
  • Operating temperature range of -65°F to +250°F
  • Features a 1/2 in. and 5/8 in. tube outside diameter configuration
Catalog No. 50-278-4613 Supplier Grainger Supplier No. 18E813
This brass extruded reducer coupling features a 45° flare design, providing a reliable metal-to-metal seal for high-pressure fluid and gas handling applications. Engineered for safety and dependability, this connector is designed for use with copper, aluminum, brass, and welded steel hydraulic tubing that can be flared. It offers an efficient alternative to soldered joints in environments where open flames are impractical.
  • Constructed from durable brass
  • Designed to meet SAE J512 and SAE J513 standards
  • Suitable for use with water, air, inert and non-combustible gases, flammable liquids, and Freon
  • Operating temperature range of -65°F to +250°F
  • Features a 1/2 in. and 5/8 in. tube outside diameter configuration